5-(4-Methoxyphenyl)thiophene-2-carboxaldehyde, 99%
5-(4-Methoxyphenyl)thiophene-2-carboxaldehyde, 99%
5-(4-Methoxyphenyl)thiophene-2-carboxaldehyde, 99%
5-(4-Methoxyphenyl)thiophene-2-carboxaldehyde, 99%
Thermo Scientific Chemicals

5-(4-Methoxyphenyl)thiophene-2-carboxaldehyde, 99%

CAS: 38401-67-1 | C13H12O2S | 232.30 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H50483.03
also known as H50483-03
Price (USD)
86.65
Special offer
Online exclusive
Ends: 30-Jun-2026
102.00
Save 15.35 (15%)
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)
86.65
Special offer
Online exclusive
Ends: 30-Jun-2026
102.00
Save 15.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS38401-67-1
IUPAC Name5-(4-methoxyphenyl)thiophene-2-carbaldehyde
Molecular FormulaC12H10O2S
InChI KeyODLOZSNXTQAWGQ-UHFFFAOYSA-N
SMILESCOC1=CC=C(C=C1)C1=CC=C(S1)C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale brown
FormCrystals or crystalline powder or powder or solid
Assay (HPLC)≥98.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only