4-Chloro-N-methylpyridine-2-carboxamide, 97%
4-Chloro-N-methylpyridine-2-carboxamide, 97%
4-Chloro-N-methylpyridine-2-carboxamide, 97%
Thermo Scientific Chemicals

4-Chloro-N-methylpyridine-2-carboxamide, 97%

CAS: 220000-87-3 | C7H7ClN2O | 170.60 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H50535.03
also known as H50535-03
Price (USD)
225.65
Online Exclusive
250.00
Save 24.35 (10%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
225.65
Online Exclusive
250.00
Save 24.35 (10%)
Each
Chemical Identifiers
CAS220000-87-3
IUPAC Name4-chloro-N-methylpyridine-2-carboxamide
Molecular FormulaC7H7ClN2O
InChI KeyBGVBBMZMEKXUTR-UHFFFAOYSA-N
SMILESCNC(=O)C1=NC=CC(Cl)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only