1-(4-Bromophenyl)pyrrolidin-2-one, 98%
1-(4-Bromophenyl)pyrrolidin-2-one, 98%
1-(4-Bromophenyl)pyrrolidin-2-one, 98%
Thermo Scientific Chemicals

1-(4-Bromophenyl)pyrrolidin-2-one, 98%

CAS: 7661-32-7 | C10H10BrNO | 240.1 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H50563.03
also known as H50563-03
Price (USD)
100.65
Online Exclusive
112.00
Save 11.35 (10%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
100.65
Online Exclusive
112.00
Save 11.35 (10%)
Each
Specifications
CAS7661-32-7
Chemical Name or Material1-(4-Bromophenyl)pyrrolidin-2-one
Formula Weight240.1
IUPAC Name1-(4-bromophenyl)pyrrolidin-2-one
InChI KeyYINFEFUSAQRZGG-UHFFFAOYSA-N
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only