Ethyl 2-bromophenylacetate, 98%
Ethyl 2-bromophenylacetate, 98%
Ethyl 2-bromophenylacetate, 98%
Thermo Scientific Chemicals

Ethyl 2-bromophenylacetate, 98%

CAS: 2178-24-7 | C10H11BrO2 | 243.1 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H50630.03
also known as H50630-03
Price (USD)
109.65
Special offer
Online exclusive
Ends: 30-Jun-2026
129.00
Save 19.35 (15%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
109.65
Special offer
Online exclusive
Ends: 30-Jun-2026
129.00
Save 19.35 (15%)
Each
Specifications
CAS2178-24-7
Chemical Name or MaterialEthyl 2-bromophenylacetate
Formula Weight243.1
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only