3-Methyl-5-phenyl-1H-pyrazole, Thermo Scientific Chemicals
3-Methyl-5-phenyl-1H-pyrazole, Thermo Scientific Chemicals
3-Methyl-5-phenyl-1H-pyrazole, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Methyl-5-phenyl-1H-pyrazole, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
H50650.03
also known as H50650-03
1 g
Catalog number H50650.03
also known as H50650-03
Price (USD)
123.65
Online Exclusive
137.00
Save 13.35 (10%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
123.65
Online Exclusive
137.00
Save 13.35 (10%)
Each
Specifications
CAS3347-62-4
Chemical Name or Material3-Methyl-5-phenyl-1H-pyrazole
EINECS Number222-099-1
Formula Weight158.2
IUPAC Name5-methyl-3-phenyl-1H-pyrazole
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only