2-(1H-Pyrazol-3-yl)pyridine, 98%
2-(1H-Pyrazol-3-yl)pyridine, 98%
2-(1H-Pyrazol-3-yl)pyridine, 98%
Thermo Scientific Chemicals

2-(1H-Pyrazol-3-yl)pyridine, 98%

CAS: 75415-03-1 | C8H7N3 | 145.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H50670.06
also known as H50670-06
Price (USD)
351.65
Online Exclusive
390.00
Save 38.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
351.65
Online Exclusive
390.00
Save 38.35 (10%)
Each
Chemical Identifiers
CAS75415-03-1
IUPAC Name2-(1H-pyrazol-5-yl)pyridine
Molecular FormulaC8H7N3
InChI KeyHKEWOTUTAYJWQJ-UHFFFAOYSA-N
SMILESN1N=CC=C1C1=CC=CC=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)98%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Notes
Incompatible with strong oxidizing agents, strong acids and bases.
RUO – Research Use Only

General References:

  1. Zhang, Q.; Chen, J.; Wu, X. Y.; Chen, X. L.; Yu, R.; Lu, C. Z.; Affiliations, S. Outstanding blue delayed fluorescence and significant processing stability of cuprous complexes with functional pyridine-pyrazolate diimine ligands. Dalton Trans. 2015, 44 (15), 6706-6710.
  2. Cao, H. T.; Shan, G. G.; Yin, Y. M.; Sun, H. Z.; Wu, Y.; Xie, W. F.; Su, Z. M. Manipulating efficiencies through modification of N-heterocyclic phenyltriazole ligands for blue iridium(III) complexes. Dyes Pigm 2015, 133, 655-663.