2-[4-(1,3,4-Oxadiazol-2-yl)phenoxy]acetamidoxime, 97%
2-[4-(1,3,4-Oxadiazol-2-yl)phenoxy]acetamidoxime, 97%
2-[4-(1,3,4-Oxadiazol-2-yl)phenoxy]acetamidoxime, 97%
2-[4-(1,3,4-Oxadiazol-2-yl)phenoxy]acetamidoxime, 97%
Thermo Scientific Chemicals

2-[4-(1,3,4-Oxadiazol-2-yl)phenoxy]acetamidoxime, 97%

CAS: 258521-20-9 | C10H10N4O3 | 234.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H50673.MD
also known as H50673-MD
Price (USD)
66.00
Each
Add to cart
Quantity:
250 mg
Request bulk or custom format
Price (USD)
66.00
Each
Add to cart
Specifications
CAS258521-20-9
Chemical Name or Material2-[4-(1,3,4-Oxadiazol-2-yl)phenoxy]acetamidoxime
Formula Weight234.22
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only