2-Phenoxypropylamine, 98%
2-Phenoxypropylamine, 98%
2-Phenoxypropylamine, 98%
Thermo Scientific Chemicals

2-Phenoxypropylamine, 98%

CAS: 6437-49-6 | C9H14ClNO | 187.667 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H50720.03
also known as H50720-03
Price (USD)
295.65
Online Exclusive
328.00
Save 32.35 (10%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
295.65
Online Exclusive
328.00
Save 32.35 (10%)
Each
Chemical Identifiers
CAS6437-49-6
IUPAC Name2-phenoxypropan-1-amine hydrochloride
Molecular FormulaC9H14ClNO
InChI KeyCGOBBGUREQJPPH-UHFFFAOYNA-N
SMILESCl.CC(CN)OC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)97.5% min
2-Phenoxypropylamine is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Phenoxypropylamine is used as pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Air sensitive. Incompatible with oxidizing agents and carbon dioxide.
RUO – Research Use Only