1-(5-Chloro-2-pyridyl)piperazine, 99%
1-(5-Chloro-2-pyridyl)piperazine, 99%
1-(5-Chloro-2-pyridyl)piperazine, 99%
Thermo Scientific Chemicals

1-(5-Chloro-2-pyridyl)piperazine, 99%

CAS: 87394-65-8 | C9H12ClN3 | 197.666 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H50862.MD
also known as H50862-MD
Price (USD)
61.65
Special offer
Online exclusive
Ends: 30-Jun-2026
71.80
Save 10.15 (14%)
Each
Quantity:
250 mg
Request bulk or custom format
Price (USD)
61.65
Special offer
Online exclusive
Ends: 30-Jun-2026
71.80
Save 10.15 (14%)
Each
Specifications
CAS87394-65-8
Chemical Name or Material1-(5-Chloro-2-pyridyl)piperazine
Formula Weight197.67
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only