3-Phenoxypiperidine, 97%
3-Phenoxypiperidine, 97%
3-Phenoxypiperidine, 97%
Thermo Scientific Chemicals

3-Phenoxypiperidine, 97%

CAS: 151666-08-9 | C11H15NO | 177.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H50912.03
also known as H50912-03
Price (USD)
266.65
Online Exclusive
296.00
Save 29.35 (10%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
266.65
Online Exclusive
296.00
Save 29.35 (10%)
Each
Chemical Identifiers
CAS151666-08-9
IUPAC Name3-phenoxypiperidine
Molecular FormulaC11H15NO
InChI KeyQDPNIJKUTWDGMV-UHFFFAOYNA-N
SMILESC1CNCC(C1)OC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.5405-1.5455 @ 20?C (or as melt)
Appearance (Color)White or clear colorless
FormFused solid or liquid
Assay (GC)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only