1-(2-Fluorophenyl)-2-propylamine, 97%
1-(2-Fluorophenyl)-2-propylamine, 97%
1-(2-Fluorophenyl)-2-propylamine, 97%
Thermo Scientific Chemicals

1-(2-Fluorophenyl)-2-propylamine, 97%

CAS: 1716-60-5 | C9H12FN | 153.20 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H50999.03
also known as H50999-03
Price (USD)
228.65
Special offer
Online exclusive
Ends: 30-Jun-2026
269.00
Save 40.35 (15%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
228.65
Special offer
Online exclusive
Ends: 30-Jun-2026
269.00
Save 40.35 (15%)
Each
Specifications
CAS1716-60-5
Chemical Name or Material1-(2-Fluorophenyl)-2-propylamine
Formula Weight153.2
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
View more
It is employed as intermediate for pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as intermediate for pharmaceutical.

Solubility
Slightly Soluble (2.0 g/L) (25°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only

General References:

  1. Yukiko Nakazono; Kenji Tsujikawa; Kenji Kuwayama; Tatsuyuki Kanamori; Yuko T. Iwata; Kazuna Miyamoto; Fumiyo Kasuya; Hiroyuki Inoue. Differentiation of regioisomeric fluoroamphetamine analogs by gas chromatography-mass spectrometry and liquid chromatography-tandem mass spectrometry. Forensic Toxicology. 2013, 31(2), 241-250.
  2. Ryouichi Nonaka; Fumiko Nagai; Akio Ogata; Kanako Satoh. In Vitro Screening of Psychoactive Drugs by [35S]GTPγS Binding in Rat Brain Membranes. Biological and Pharmaceutical Bulletin. 2007, 30(12), 2328-2333.