4-(Methoxymethyl)piperidine hydrochloride
4-(Methoxymethyl)piperidine hydrochloride
4-(Methoxymethyl)piperidine hydrochloride
Thermo Scientific Chemicals

4-(Methoxymethyl)piperidine hydrochloride

CAS: 916317-00-5 | C7H16ClNO | 165.66 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H51020.MD
also known as H51020-MD
Price (USD)
75.65
Special offer
Online exclusive
Ends: 30-Jun-2026
89.40
Save 13.75 (15%)
Each
Quantity:
250 mg
Request bulk or custom format
Price (USD)
75.65
Special offer
Online exclusive
Ends: 30-Jun-2026
89.40
Save 13.75 (15%)
Each
Chemical Identifiers
CAS916317-00-5
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or crystalline powder or powder
Assay from Suppliers CofA≥97.5%
Appearance (Color)White to pale yellow
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only