2-Chloro-5,5-dimethyl-1,3-cyclohexanedione, 98%
2-Chloro-5,5-dimethyl-1,3-cyclohexanedione, 98%
2-Chloro-5,5-dimethyl-1,3-cyclohexanedione, 98%
Thermo Scientific Chemicals

2-Chloro-5,5-dimethyl-1,3-cyclohexanedione, 98%

CAS: 7298-89-7 | C8H11ClO2 | 174.624 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H51035.MD
also known as H51035-MD
Price (USD)
75.65
Online Exclusive
84.10
Save 8.45 (10%)
Each
Quantity:
250 mg
Request bulk or custom format
Price (USD)
75.65
Online Exclusive
84.10
Save 8.45 (10%)
Each
Chemical Identifiers
CAS7298-89-7
IUPAC Name2-chloro-5,5-dimethylcyclohexane-1,3-dione
Molecular FormulaC8H11ClO2
InChI KeyVOBIHUAWDXUCPH-UHFFFAOYSA-N
SMILESCC1(C)CC(=O)C(Cl)C(=O)C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)97.5% min
2-Chloro-5,5-dimethyl-1,3-cyclohexanedione is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chloro-5,5-dimethyl-1,3-cyclohexanedione is used as pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Moisture sensitive. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Saied Saeed Hosseiny Davarani; Davood Nematollahi; Mojtaba Shamsipur; Nahid Mashkouri Najafi; Leila Masoumi; Somayyeh Ramyar. Electrochemical oxidation of 2,3-dimethylhydroquinone in the presence of 1,3-dicarbonyl compounds. J. Org. Chem. 2006, 71, (5),2139-2142
  2. Saied Saeed Hosseiny Davarani; Nahid Mashkouri Najafi; Somayyeh Ramyar; Leila Masoumi; Mojtaba Shamsipur. An improved electrochemical method for the synthesis of some benzofuran derivatives. Chemical and Pharmaceutical Bulletin. 2006, 54, (7),959-962