1-Vinylnaphthalene, 95%, stab. with 4-tert-butylcatechol
1-Vinylnaphthalene, 95%, stab. with 4-tert-butylcatechol
1-Vinylnaphthalene, 95%, stab. with 4-tert-butylcatechol
Thermo Scientific Chemicals

1-Vinylnaphthalene, 95%, stab. with 4-tert-butylcatechol

CAS: 826-74-4 | C12H10 | 154.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H51043.06
Price (USD)
261.65
Online Exclusive
290.00
Save 28.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
261.65
Online Exclusive
290.00
Save 28.35 (10%)
Each
Chemical Identifiers
CAS826-74-4
IUPAC Name1-ethenylnaphthalene
Molecular FormulaC12H10
InChI KeyIGGDKDTUCAWDAN-UHFFFAOYSA-N
SMILESC=CC1=C2C=CC=CC2=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)>94.0%
1-Vinylnaphthalene is used as a precursor of polynuclear aromatic hydrocarbons in tobacco smoke. It is also used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Vinylnaphthalene is used as a precursor of polynuclear aromatic hydrocarbons in tobacco smoke. It is also used as pharmaceutical intermediate.

Solubility
Soluble in ethyl acetate, hexane and methanol.

Notes
Moisture sensitive. Incompatible with oxidizing agents, heat, light, ultraviolet radiation and free radical initiators.
RUO – Research Use Only

General References:

  1. Robert F.Cozzens; Robert B.Fox. Intramolecular triplet energy transfer in poly(1-vinylnaphthalene). J. Chem. Phys. 1969, 50, (4),1532
  2. C.E.Hoyle; T.L.Nemzek; A.Mar; J.E.Guillet. Time resolved fluorescence studies of Poly(N-vinylcarbazole), Poly(1-vinylnaphthalene), and 1,3-Bis(N-carbazolyl)propane. Macromolecules. 1978, 11, (2),429-431