4-Amino-1-(2-pyridyl)piperidine hydrochloride, 97%
4-Amino-1-(2-pyridyl)piperidine hydrochloride, 97%
4-Amino-1-(2-pyridyl)piperidine hydrochloride, 97%
4-Amino-1-(2-pyridyl)piperidine hydrochloride, 97%
Thermo Scientific Chemicals

4-Amino-1-(2-pyridyl)piperidine hydrochloride, 97%

CAS: 77145-39-2 | C10H16ClN3 | 213.709 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H51108.MD
also known as H51108-MD
Price (USD)
55.65
Special offer
Online exclusive
Ends: 30-Jun-2026
64.80
Save 9.15 (14%)
Each
Add to cart
Quantity:
250 mg
Request bulk or custom format
Price (USD)
55.65
Special offer
Online exclusive
Ends: 30-Jun-2026
64.80
Save 9.15 (14%)
Each
Add to cart
Specifications
CAS77145-39-2
Chemical Name or Material4-Amino-1-(2-pyridyl)piperidine hydrochloride
Formula Weight213.71
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
View more
4-Amino-1-(2-pyridyl)piperidine hydrochloride is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Amino-1-(2-pyridyl)piperidine hydrochloride is used as pharmaceutical intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only
A.Michael Crider; Randall Lamey; Heinz G.Floss; John M.Cassady; William J.Bradner. Synthesis of nitrosourea derivatives of pyridine and piperidine as potential anticancer agents. J. Med. Chem. 1980, 23, (8),848-851