3-Iodobenzeneboronic acid, 97%
3-Iodobenzeneboronic acid, 97%
3-Iodobenzeneboronic acid, 97%
3-Iodobenzeneboronic acid, 97%
Thermo Scientific Chemicals

3-Iodobenzeneboronic acid, 97%

CAS: 221037-98-5 | C6H6BIO2 | 247.826 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H51112.03
also known as H51112-03
Price (USD)
29.00
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)
29.00
Each
Add to cart
Specifications
CAS221037-98-5
Chemical Name or Material3-Iodobenzeneboronic acid
Formula Weight247.83
Health Hazard 1H302-H315-H319-H335
Health Hazard 2GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only