1,3-Difluoroacetone, 98%
1,3-Difluoroacetone, 98%
1,3-Difluoroacetone, 98%
Thermo Scientific Chemicals

1,3-Difluoroacetone, 98%

CAS: 453-14-5 | C3H4F2O | 94.061 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H51145.03
also known as H51145-03
Price (USD)
92.65
Special offer
Online exclusive
Ends: 30-Jun-2026
109.00
Save 16.35 (15%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
92.65
Special offer
Online exclusive
Ends: 30-Jun-2026
109.00
Save 16.35 (15%)
Each
Specifications
CAS453-14-5
Chemical Name or Material1,3-Difluoroacetone
DOT InformationTransport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC LIQUIDS, FLAMMABLE, ORGANIC, N.O.S.
Formula Weight94.06
Health Hazard 1H226-H301+H311+H331
View more
1,3-Difluoroacetone is used as pharmaceutical intermediate and for chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3-Difluoroacetone is used as pharmaceutical intermediate and for chemical research.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and bases.
RUO – Research Use Only
David J.Finnigan; Charles W.Gillies; Richard D.Suenram; E.Bright Wilson; Hasse Karlsson. Microwave determination of the conformation and dipole moment of 1,3-difluoroacetone. Journal of Molecular Spectroscopy. 1975, 57, (3),363-376