cis-1-Ethoxy-2-(tri-n-butylstannyl)ethylene, 94%
cis-1-Ethoxy-2-(tri-n-butylstannyl)ethylene, 94%
cis-1-Ethoxy-2-(tri-n-butylstannyl)ethylene, 94%
Thermo Scientific Chemicals

cis-1-Ethoxy-2-(tri-n-butylstannyl)ethylene, 94%

CAS: 64724-29-4 | C16H34OSn | 361.157 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number H51258.03
also known as H51258-03
Price (USD)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS64724-29-4
IUPAC Nametributyl(2-ethoxyethenyl)stannane
Molecular FormulaC16H34OSn
InChI KeyWARKYKQCOXTIAO-UHFFFAOYSA-N
SMILESCCCC[Sn](CCCC)(CCCC)C=COCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless
FormLiquid
Assay (GC)>92.5%
cis-1-Ethoxy-2-(tri-n-butylstannyl)ethylene is widely used in pharmaceutical research and as reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
cis-1-Ethoxy-2-(tri-n-butylstannyl)ethylene is widely used in pharmaceutical research and as reagent.

Solubility
Soluble in water.

Notes
cis-1-Ethoxy-2-(tri-n-butylstannyl)ethylene is to be stored in a cool, dry place and is to be sealed properly.
RUO – Research Use Only
Suzuki, M.; Iwasaki, H.; Fujikawa, Y.; Kitahara, M.; Sakashita, M.; Sakoda, R. Synthesis and biological evaluations of quinoline-based HMG-CoA reductase inhibitors. Bioorgan. Med. Chem. 2001, 9 (10), 2727-2743.