4-(Tri-n-butylstannyl)pyridine, 96%
4-(Tri-n-butylstannyl)pyridine, 96%
4-(Tri-n-butylstannyl)pyridine, 96%
Thermo Scientific Chemicals

4-(Tri-n-butylstannyl)pyridine, 96%

CAS: 124252-41-1 | C17H31NSn | 368.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number H51435.06
also known as H51435-06
Price (USD)
-
Quantity:
5 g
Request bulk or custom format
Specifications
CAS124252-41-1
Chemical Name or Material4-(Tri-n-butylstannyl)pyridine
DOT InformationTransport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOTIN COMPOUNDS, LIQUID, N.O.S.
Formula Weight368.15
Health Hazard 1H301-H312-H315-H319-H372
View more
4-(Tri-n-butylstannyl)pyridine is used as organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(Tri-n-butylstannyl)pyridine is used as organic chemical synthesis intermediate.

Solubility
Not miscible in water. Soluble in ether, hexane and tetrahydrofuran.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only
Bruce A.Reinhardt; Lawrence L.Brott; Stephen J.Clarson; Ann G.Dillard; Jayprakash C.Bhatt; Ramamurthi Kannan; Lixiang Yuan; Guang S.He; Paras N.Prasad. Highly active two-photon dyes:  Design, synthesis, and characterization toward application. Chem. Mater. 1998, 10, (7),1863-1874