3-Methyl-2-(tri-n-butylstannyl)pyridine, 96%
3-Methyl-2-(tri-n-butylstannyl)pyridine, 96%
3-Methyl-2-(tri-n-butylstannyl)pyridine, 96%
Thermo Scientific Chemicals

3-Methyl-2-(tri-n-butylstannyl)pyridine, 96%

CAS: 259807-97-1 | C18H33NSn | 382.179 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number H51540.MD
also known as H51540-MD
Price (USD)
-
Quantity:
250 mg
Request bulk or custom format
Specifications
CAS259807-97-1
Chemical Name or Material3-Methyl-2-(tri-n-butylstannyl)pyridine
DOT InformationTransport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOTIN COMPOUNDS, LIQUID, N.O.S.
Formula Weight382.16
Health Hazard 1H301-H312-H315-H319-H372
View more
3-Methyl-2-(tri-n-butylstannyl)pyridine is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Methyl-2-(tri-n-butylstannyl)pyridine is used as pharmaceutical intermediate.

Solubility
Sparingly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and heat.
RUO – Research Use Only
Angela Gurski Birchler; Fuqiang Liu; Lanny S.Liebeskind. Synthesis of .alpha.-pyridone-based azaheteroaromatics by intramolecular vinylketene cyclizations onto the C:N bond of nitrogen heteroaromatics. J. Org. Chem. 1994, 59, (25),7737-7745