1,4-Benzenediboronic acid bis(pinacol) ester, 97%
1,4-Benzenediboronic acid bis(pinacol) ester, 97%
1,4-Benzenediboronic acid bis(pinacol) ester, 97%
Thermo Scientific Chemicals

1,4-Benzenediboronic acid bis(pinacol) ester, 97%

CAS: 99770-93-1 | C18H28B2O4 | 330.038 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H51707.03
also known as H51707-03
Price (USD)
55.65
Online Exclusive
61.60
Save 5.95 (10%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
55.65
Online Exclusive
61.60
Save 5.95 (10%)
Each
Specifications
CAS99770-93-1
Chemical Name or Material1,4-Benzenediboronic acid bis(pinacol) ester
Formula Weight330.03
Health Hazard 1H315-H319-H335
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only