Specifications
CAS5346-38-3
Chemical Name or MaterialPyridine-2-thiocarboxamide
Formula Weight138.19
Health Hazard 1H302-H315-H317-H318-H335
Health Hazard 2GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
View more