Thiophene-2,5-diboronic acid bis(pinacol) ester, 97%
Thiophene-2,5-diboronic acid bis(pinacol) ester, 97%
Thiophene-2,5-diboronic acid bis(pinacol) ester, 97%
Thiophene-2,5-diboronic acid bis(pinacol) ester, 97%
Thermo Scientific Chemicals

Thiophene-2,5-diboronic acid bis(pinacol) ester, 97%

CAS: 175361-81-6 | C16H26B2O4S | 336.06 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H51725.03
also known as H51725-03
Price (USD)
74.65
Online Exclusive
83.30
Save 8.65 (10%)
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)
74.65
Online Exclusive
83.30
Save 8.65 (10%)
Each
Add to cart
Specifications
CAS175361-81-6
Chemical Name or MaterialThiophene-2,5-diboronic acid bis(pinacol) ester
Formula Weight336.06
IUPAC Name4,4,5,5-tetramethyl-2-[5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thiophen-2-yl]-1,3,2-dioxaborolane
InChI KeyAOJXAKMKFDBHHT-UHFFFAOYSA-N
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only