Disuccinimidyl suberate, 97%
Disuccinimidyl suberate, 97%
Disuccinimidyl suberate, 97%
Thermo Scientific Chemicals

Disuccinimidyl suberate, 97%

CAS: 68528-80-3 | C16H20N2O8 | 368.342 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mg
1 g
Catalog number H51771.03
also known as H51771-03
Price (USD)
124.65
Special offer
Online exclusive
Ends: 28-Dec-2026
147.00
Save 22.35
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
124.65
Special offer
Online exclusive
Ends: 28-Dec-2026
147.00
Save 22.35
Each
Specifications
CAS68528-80-3
Chemical Name or MaterialDisuccinimidyl suberate
Formula Weight368.34
Health Hazard 1H315-H319-H335
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
View more
Disuccinimidyl suberate is a homobifunctional amine-reactive crosslinker. It is usually coupled to molecules containing primary amines via amide bonds buffered at pH 7.5 (6.5-8.5). It incorporates an 8-atom spacer arm.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Disuccinimidyl suberate is a homobifunctional amine-reactive crosslinker. It is usually coupled to molecules containing primary amines via amide bonds buffered at pH 7.5 (6.5-8.5). It incorporates an 8-atom spacer arm.

Solubility
Insoluble in water. Soluble in chloroform: 50 mg/mL.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. J.J.Leary; D.J.Brigati; D.C.Ward. Rapid and sensitive colorimetric method for visualizing biotin-labeled DNA probes hybridized to DNA or RNA immobilized on nitrocellulose: Bio-blots. Current Issue. 1983, 80, (13),4045-4049
  2. S.Cheifetz; T.Bellon; C.Cales; S.Vera; C.Bernabeu; J.Massague; M.Letarte. Endoglin is a component of the transforming growth factor-beta receptor system in human endothelial cells. The Journal of Biological Chemistry. 1992, 267, (27),19027-19030