Specifications
CAS2208-59-5
Chemical Name or Material2-(4-Pyridyl)benzimidazole
Formula Weight195.22
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
View more