Benzene-1,4-dithiocarboxamide, 97%
Benzene-1,4-dithiocarboxamide, 97%
Benzene-1,4-dithiocarboxamide, 97%
Thermo Scientific Chemicals

Benzene-1,4-dithiocarboxamide, 97%

CAS: 13363-51-4 | C8H8N2S2 | 196.286 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H51857.03
also known as H51857-03
Price (USD)
72.90
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
72.90
Each
Specifications
CAS13363-51-4
Chemical Name or MaterialBenzene-1,4-dithiocarboxamide
DOT InformationTransport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
Formula Weight196.28
Health Hazard 1H301
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only