2-Chloroimidazole, 97%
2-Chloroimidazole, 97%
2-Chloroimidazole, 97%
Thermo Scientific Chemicals

2-Chloroimidazole, 97%

CAS: 16265-04-6 | C3H3ClN2 | 102.52 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H51863.03
also known as H51863-03
Price (USD)
249.65
Special offer
Online exclusive
Ends: 30-Jun-2026
293.00
Save 43.35 (15%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
249.65
Special offer
Online exclusive
Ends: 30-Jun-2026
293.00
Save 43.35 (15%)
Each
Chemical Identifiers
CAS16265-04-6
SpecificationsSpecification SheetSpecification Sheet
FormSolid
Appearance (Color)Off-white
Assay (unspecified)>96.0%
2-Chloroimidazole is used in pharmaceuticals, drug candidates, ligands for transition metal catalysts and other molecular functional materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chloroimidazole is used in pharmaceuticals, drug candidates, ligands for transition metal catalysts and other molecular functional materials.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Neelakandha S.Mani; Jill A.Jablonowski;Todd K.Jones. A scalable synthesis of a histamine H3 receptor antagonist. J. Org. Chem. 2004, 69, (23),8115-8117
  2. Y.Takeuchi; Kenneth L.Kirk; Louis A.Cohen. Acid-catalyzed isotope exchange of ring hydrogens in fluoroimidazoles. J. Org. Chem. 1979, 44, (24),4240-4242