2-Chloro-1-methylimidazole, 97+%
2-Chloro-1-methylimidazole, 97+%
2-Chloro-1-methylimidazole, 97+%
Thermo Scientific Chemicals

2-Chloro-1-methylimidazole, 97+%

CAS: 253453-91-7 | C4H5ClN2 | 116.55 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H51867.03
also known as H51867-03
Price (USD)
176.65
Online Exclusive
196.00
Save 19.35 (10%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
176.65
Online Exclusive
196.00
Save 19.35 (10%)
Each
Chemical Identifiers
CAS253453-91-7
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)≥97.0% (GC)
Proton NMRConforms to structure
Appearance (Color)Colorless to pale brown
FormLiquid or viscous liquid
Assay≥97.0% (GC)
2-Chloro-1-methylimidazole is used in pharmaceuticals, drug candidates, ligands for transition metal catalysts and other molecular functional materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chloro-1-methylimidazole is used in pharmaceuticals, drug candidates, ligands for transition metal catalysts and other molecular functional materials.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only
E.V.Aleksandrova; A.N.Kravchenko; P.M.Kochergin. Methods for the synthesis of haloimidazoles (review). Chemistry of Heterocyclic Compounds. 2011, 46, (11),1295-1317