4-(Diphenylamino)benzeneboronic acid pinacol ester, 95%
4-(Diphenylamino)benzeneboronic acid pinacol ester, 95%
4-(Diphenylamino)benzeneboronic acid pinacol ester, 95%
Thermo Scientific Chemicals

4-(Diphenylamino)benzeneboronic acid pinacol ester, 95%

CAS: 267221-88-5 | C24H26BNO2 | 371.29 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H51946.06
also known as H51946-06
Price (USD)
223.65
Online Exclusive
248.00
Save 24.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
223.65
Online Exclusive
248.00
Save 24.35 (10%)
Each
Specifications
Beilstein10571706
CAS267221-88-5
Chemical Name or Material4-(Diphenylamino)benzeneboronic acid pinacol ester
Formula Weight371.29
IUPAC NameN,N-diphenyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only