9,9-Dimethylfluorene-2-boronic acid pinacol ester, 95%
9,9-Dimethylfluorene-2-boronic acid pinacol ester, 95%
9,9-Dimethylfluorene-2-boronic acid pinacol ester, 95%
Thermo Scientific Chemicals

9,9-Dimethylfluorene-2-boronic acid pinacol ester, 95%

CAS: 569343-09-5 | C21H25BO2 | 320.239 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H51950.06
also known as H51950-06
Price (USD)
178.65
Online Exclusive
198.00
Save 19.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
178.65
Online Exclusive
198.00
Save 19.35 (10%)
Each
Specifications
CAS569343-09-5
Chemical Name or Material9,9-Dimethylfluorene-2-boronic acid pinacol ester
Formula Weight320.24
IUPAC Name2-(9,9-dimethylfluoren-2-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
InChI KeyDAZFRJAIIUPRQZ-UHFFFAOYSA-N
View more
It is employed in the synthesis and photo physical properties of 1,2-diphenylindolizine derivatives: fluorescent blue-emitting materials for organic light-emitting device.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed in the synthesis and photo physical properties of 1,2-diphenylindolizine derivatives: fluorescent blue-emitting materials for organic light-emitting device.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

General References:

  1. Andrea Cappelli, et al. Highly emissive supramolecular assemblies based on π-stacked polybenzofulvene hosts and a benzothiadiazole guest.J. Mater. Chem. C.,2014,2(37), 7897-7905.
  2. Yeong Ran Song, et al. Synthesis and photo physical properties of 1,2-diphenylindolizine derivatives: fluorescent blue-emitting materials for organic light-emitting device.Luminescence.,2015,