9,9-Di-n-octylfluorene-2-boronic acid pinacol ester, 95%, Thermo Scientific Chemicals
9,9-Di-n-octylfluorene-2-boronic acid pinacol ester, 95%, Thermo Scientific Chemicals
9,9-Di-n-octylfluorene-2-boronic acid pinacol ester, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

9,9-Di-n-octylfluorene-2-boronic acid pinacol ester, 95%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
H51953.06
also known as H51953-06
5 g
Catalog number H51953.06
also known as H51953-06
Price (USD)
214.65
Online Exclusive
238.00
Save 23.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
214.65
Online Exclusive
238.00
Save 23.35 (10%)
Each
Specifications
Beilstein8661191
CAS302554-81-0
Chemical Name or Material9,9-Di-n-octylfluorene-2-boronic acid pinacol ester
Formula Weight516.61
IUPAC Name2-(9,9-dioctylfluoren-2-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only