(R)-N-Boc-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid, 98%
(R)-N-Boc-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid, 98%
(R)-N-Boc-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid, 98%
Thermo Scientific Chemicals

(R)-N-Boc-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid, 98%

CAS: 115962-35-1 | C15H18NO4 | 276.31 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H51965.06
also known as H51965-06
Price (USD)
252.65
Online Exclusive
280.00
Save 27.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
252.65
Online Exclusive
280.00
Save 27.35 (10%)
Each
Chemical Identifiers
CAS115962-35-1
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)97.5% min
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only