N-Fmoc-3-(1-naphthyl)-L-alanine, 98%
N-Fmoc-3-(1-naphthyl)-L-alanine, 98%
N-Fmoc-3-(1-naphthyl)-L-alanine, 98%
Thermo Scientific Chemicals

N-Fmoc-3-(1-naphthyl)-L-alanine, 98%

CAS: 96402-49-2 | C28H23NO4 | 437.50 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H51967.03
also known as H51967-03
Price (USD)
160.65
Special offer
Online exclusive
Ends: 30-Jun-2026
189.00
Save 28.35 (15%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
160.65
Special offer
Online exclusive
Ends: 30-Jun-2026
189.00
Save 28.35 (15%)
Each
Specifications
Beilstein4829118
CAS96402-49-2
Chemical Name or MaterialN-Fmoc-3-(1-naphthyl)-L-alanine
Formula Weight437.49
IUPAC Name(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-naphthalen-1-ylpropanoic acid
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only