(R)-3-(Boc-amino)-3-phenylpropionic acid, 95%, Thermo Scientific Chemicals
(R)-3-(Boc-amino)-3-phenylpropionic acid, 95%, Thermo Scientific Chemicals
(R)-3-(Boc-amino)-3-phenylpropionic acid, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

(R)-3-(Boc-amino)-3-phenylpropionic acid, 95%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
H51984.03
also known as H51984-03
1 g
Catalog number H51984.03
also known as H51984-03
Price (USD)
206.65
Special offer
Online exclusive
Ends: 30-Jun-2026
243.00
Save 36.35 (15%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
206.65
Special offer
Online exclusive
Ends: 30-Jun-2026
243.00
Save 36.35 (15%)
Each
Specifications
Beilstein5404389
CAS161024-80-2
Chemical Name or Material(R)-3-(Boc-amino)-3-phenylpropionic acid
Formula Weight265.31
IUPAC Name(3R)-3-[(2-methylpropan-2-yl)oxycarbonylamino]-3-phenylpropanoic acid
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only