N-Boc-4-nitro-L-phenylalanine, 95%
N-Boc-4-nitro-L-phenylalanine, 95%
N-Boc-4-nitro-L-phenylalanine, 95%
Thermo Scientific Chemicals

N-Boc-4-nitro-L-phenylalanine, 95%

CAS: 33305-77-0 | C14H18N2O6 | 310.306 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H51986.03
also known as H51986-03
Price (USD)
42.65
Special offer
Online exclusive
Ends: 30-Jun-2026
50.00
Save 7.35 (15%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
42.65
Special offer
Online exclusive
Ends: 30-Jun-2026
50.00
Save 7.35 (15%)
Each
Specifications
Beilstein2820671
CAS33305-77-0
Chemical Name or MaterialN-Boc-4-nitro-L-phenylalanine
EINECS Number251-450-1
Formula Weight310.31
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only