(S)-4-(Boc-amino)-2-(Fmoc-amino)butyric acid, 96%
(S)-4-(Boc-amino)-2-(Fmoc-amino)butyric acid, 96%
(S)-4-(Boc-amino)-2-(Fmoc-amino)butyric acid, 96%
Thermo Scientific Chemicals

(S)-4-(Boc-amino)-2-(Fmoc-amino)butyric acid, 96%

CAS: 125238-99-5 | C24H28N2O6 | 440.50 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H51990.03
also known as H51990-03
Price (USD)
100.65
Online Exclusive
112.00
Save 11.35 (10%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
100.65
Online Exclusive
112.00
Save 11.35 (10%)
Each
Chemical Identifiers
CAS125238-99-5
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)95.0% min
It finds its application in combination with the Nγ-Z derivative for the synthesis of polymyxin analogs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds its application in combination with the Nγ-Z derivative for the synthesis of polymyxin analogs.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep it refrigerated.
RUO – Research Use Only

General References:

  1. Sherry M. Tsai, et al. Unanticipated differences between α- and γ-diaminobutyric acid-linked hairpin polyamide-alkylator conjugates.Nucl. Acids Res.,2007,35(1), 307-316.
  2. Kazushi Kanazawa, et al. Contribution of Each Amino Acid Residue in Polymyxin B3 to Antimicrobial and Lipopolysaccharide Binding Activity.Chem. Pharm. Bull.,2009,57(3), 240-244.