3-(4-Biphenylyl)-N-Fmoc-L-alanine, 95%
3-(4-Biphenylyl)-N-Fmoc-L-alanine, 95%
3-(4-Biphenylyl)-N-Fmoc-L-alanine, 95%
3-(4-Biphenylyl)-N-Fmoc-L-alanine, 95%
Thermo Scientific Chemicals

3-(4-Biphenylyl)-N-Fmoc-L-alanine, 95%

CAS: 199110-64-0 | C30H25NO4 | 463.533 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
5 g
Catalog number H52002.06
also known as H52002-06
Price (USD)
577.65
Special offer
Online exclusive
Ends: 30-Jun-2026
679.00
Save 101.35 (15%)
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
577.65
Special offer
Online exclusive
Ends: 30-Jun-2026
679.00
Save 101.35 (15%)
Each
Add to cart
Specifications
CAS199110-64-0
Chemical Name or Material3-(4-Biphenylyl)-N-Fmoc-L-alanine
Formula Weight463.52
IUPAC Name(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-(4-phenylphenyl)propanoic acid
InChI KeyVSGACONKQRJFGX-NDEPHWFRSA-N
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only