2-Benzyl-D-proline hydrochloride, 95%, Thermo Scientific Chemicals
2-Benzyl-D-proline hydrochloride, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Benzyl-D-proline hydrochloride, 95%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
H52019.03
also known as H52019-03
1 g
Catalog number H52019.03
also known as H52019-03
Price (USD)
425.65
Online Exclusive
473.00
Save 47.35 (10%)
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)
425.65
Online Exclusive
473.00
Save 47.35 (10%)
Each
Add to cart
Specifications
CAS1111130-04-1
Chemical Name or Material2-Benzyl-D-proline hydrochloride
Formula Weight241.72
IUPAC Name(2S)-2-benzylpyrrolidine-2-carboxylic acid;hydrochloride
InChI KeySPHJFGQFCBZKRU-UHFFFAOYNA-N
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only