N-Fmoc-3-(4-pyridyl)-L-alanine, 95%
N-Fmoc-3-(4-pyridyl)-L-alanine, 95%
N-Fmoc-3-(4-pyridyl)-L-alanine, 95%
Thermo Scientific Chemicals

N-Fmoc-3-(4-pyridyl)-L-alanine, 95%

CAS: 169555-95-7 | C23H20N2O4 | 388.423 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
5 g
Catalog number H52172.03
also known as H52172-03
Price (USD)
204.65
Online Exclusive
227.00
Save 22.35 (10%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
204.65
Online Exclusive
227.00
Save 22.35 (10%)
Each
Specifications
Beilstein7316905
CAS169555-95-7
Chemical Name or MaterialN-Fmoc-3-(4-pyridyl)-L-alanine
Formula Weight388.42
IUPAC Name(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-pyridin-4-ylpropanoic acid
View more
Fmoc-(4-pyridyl)-Ala-OH is potentially useful for solid phase peptide synthesis techniques.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Fmoc-∫-(4-pyridyl)-Ala-OH is potentially useful for solid phase peptide synthesis techniques.

Solubility
Soluble in 0.1g in 4mL DMF(clearly soluble).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents. Store at 4°C.
RUO – Research Use Only
Vommina V. Sureshbabu,; N. Narendra and G. Nagendra. Chiral N-Fmoc-β-Amino Alkyl Isonitriles Derived from Amino Acids: First Synthesis and Application in 1-Substituted Tetrazole Synthesis. J. Org. Chem., 2009, 74 (1), 153-157