N-Fmoc-3,3-diphenyl-L-alanine, 95%
N-Fmoc-3,3-diphenyl-L-alanine, 95%
N-Fmoc-3,3-diphenyl-L-alanine, 95%
Thermo Scientific Chemicals

N-Fmoc-3,3-diphenyl-L-alanine, 95%

CAS: 201484-50-6 | C30H25NO4 | 463.533 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H52176.06
also known as H52176-06
Price (USD)
549.65
Special offer
Online exclusive
Ends: 30-Jun-2026
647.00
Save 97.35 (15%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
549.65
Special offer
Online exclusive
Ends: 30-Jun-2026
647.00
Save 97.35 (15%)
Each
Chemical Identifiers
CAS201484-50-6
IUPAC Name(2S)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3,3-diphenylpropanoic acid
Molecular FormulaC30H25NO4
InChI KeyPENQOTJCVODUQU-NDEPHWFRSA-N
SMILESOC(=O)[C@@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)94.0% min
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only