N-Fmoc-3-(2-naphthyl)-L-alanine, 95%
N-Fmoc-3-(2-naphthyl)-L-alanine, 95%
N-Fmoc-3-(2-naphthyl)-L-alanine, 95%
Thermo Scientific Chemicals

N-Fmoc-3-(2-naphthyl)-L-alanine, 95%

CAS: 112883-43-9 | C28H22NO4 | 436.49 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
5 g
Catalog number H52179.03
also known as H52179-03
Price (USD)
112.65
Special offer
Online exclusive
Ends: 28-Dec-2026
132.00
Save 19.35
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
112.65
Special offer
Online exclusive
Ends: 28-Dec-2026
132.00
Save 19.35
Each
Chemical Identifiers
CAS112883-43-9
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)≥94.0%
Chiral LC≥99.5% (ee)
Water Content (Karl Fischer Titration)≤2.0%
Solution TestClear solution (0.3 g in 2mL DMF)
FormPowder
View more
N-Fmoc-3-(2-naphthyl)-L-alanine is used as organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Fmoc-3-(2-naphthyl)-L-alanine is used as organic chemical synthesis intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
RUO – Research Use Only