N-Fmoc-L-beta-glutamic acid 5-tert-butyl ester, 95%
N-Fmoc-L-beta-glutamic acid 5-tert-butyl ester, 95%
N-Fmoc-L-beta-glutamic acid 5-tert-butyl ester, 95%
N-Fmoc-L-beta-glutamic acid 5-tert-butyl ester, 95%
Thermo Scientific Chemicals

N-Fmoc-L-beta-glutamic acid 5-tert-butyl ester, 95%

CAS: 209252-17-5 | C24H27NO6 | 425.481 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H52190.03
also known as H52190-03
Price (USD)
383.65
Online Exclusive
426.00
Save 42.35 (10%)
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)
383.65
Online Exclusive
426.00
Save 42.35 (10%)
Each
Add to cart
Specifications
Beilstein7952342
CAS209252-17-5
Chemical Name or MaterialN-Fmoc-L-beta-glutamic acid 5-tert-butyl ester
Formula Weight425.48
IUPAC Name(3R)-3-(9H-fluoren-9-ylmethoxycarbonylamino)-5-[(2-methylpropan-2-yl)oxy]-5-oxopentanoic acid
View more
N-Fmoc-L-beta-glutamic acid 5-tert-butyl ester, is a protected building block used for the introduction of glutamic acid into peptides. Fmoc-beta-Glu(OtBu)-OH) was hydrolyzed with trifluoroacetic acid (TFA) and isolated with the HPLC system to give Fmoc-beta-Glu(OH)-OH.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Fmoc-L-beta-glutamic acid 5-tert-butyl ester, is a protected building block used for the introduction of glutamic acid into peptides. Fmoc-beta-Glu(OtBu)-OH) was hydrolyzed with trifluoroacetic acid (TFA) and isolated with the HPLC system to give Fmoc-beta-Glu(OH)-OH.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents. Store at 4°C.
RUO – Research Use Only
Kongzhen Hu,; Xiaolan Tang,; Ganghua Tang,; Shaobo Yao,; Baoguo Yao,; Hongliang Wang,; Dahong Nie,; Xiang Liang, Caihua Tang,; Shanzhen He.18F-FP-PEG2-β-Glu-RGD2: A Symmetric Integrin αvβ3-Targeting Radiotracer for Tumor PET Imaging. journal.pone.. 2015,