(1S,2S)-Cyclohexane-1,2-dicarboxylic acid, 98+%
(1S,2S)-Cyclohexane-1,2-dicarboxylic acid, 98+%
(1S,2S)-Cyclohexane-1,2-dicarboxylic acid, 98+%
Thermo Scientific Chemicals

(1S,2S)-Cyclohexane-1,2-dicarboxylic acid, 98+%

CAS: 21963-41-7 | C8H12O4 | 172.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H52290.03
also known as H52290-03
Price (USD)
94.00
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
94.00
Each
Specifications
CAS21963-41-7
Chemical Name or Material(1S,2S)-Cyclohexane-1,2-dicarboxylic acid
Formula Weight172.18
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
View more
A series of new C2-symmetric (1S,2S)-cyclohexane-1,2-dicarboxamides was synthesized from (1S,2S)-Cyclohexane-1,2-dicarboxylic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A series of new C2-symmetric (1S,2S)-cyclohexane-1,2-dicarboxamides was synthesized from (1S,2S)-Cyclohexane-1,2-dicarboxylic acid.

Solubility
Soluble in acetone(almost transparency).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only
Chan Zhou,; Jiaxi Xu.Synthesis of New C2-Symmetric Chiral Bisamides from (1S,2S)-Cyclohexane-1,2-dicarboxylic Acid. Helvetica Chimica Acta. 2014,97(10), 1396-1405.