(+/-)-1,4-Diazabicyclo[4.4.0]decane, 98+%
(+/-)-1,4-Diazabicyclo[4.4.0]decane, 98+%
(+/-)-1,4-Diazabicyclo[4.4.0]decane, 98+%
Thermo Scientific Chemicals

(+/-)-1,4-Diazabicyclo[4.4.0]decane, 98+%

CAS: 4430-75-5 | C8H16N2 | 140.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H52305.MD
also known as H52305-MD
Price (USD)
158.65
Online Exclusive
176.00
Save 17.35 (10%)
Each
Quantity:
250 mg
Request bulk or custom format
Price (USD)
158.65
Online Exclusive
176.00
Save 17.35 (10%)
Each
Chemical Identifiers
CAS4430-75-5
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Assay from Suppliers CofA≥98.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only