N-(2-Hydroxyethyl)formamide, 97%
N-(2-Hydroxyethyl)formamide, 97%
N-(2-Hydroxyethyl)formamide, 97%
Thermo Scientific Chemicals

N-(2-Hydroxyethyl)formamide, 97%

CAS: 693-06-1 | C3H7NO2 | 89.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H52345.03
also known as H52345-03
Price (USD)
320.65
Online Exclusive
356.00
Save 35.35 (10%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
320.65
Online Exclusive
356.00
Save 35.35 (10%)
Each
Specifications
CAS693-06-1
Chemical Name or MaterialN-(2-Hydroxyethyl)formamide
EINECS Number211-741-6
Formula Weight89.09
Health Hazard 1H315-H319-H335
View more
N-(2-Hydroxyethyl)formamide is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-(2-Hydroxyethyl)formamide is used as an organic chemical synthesis intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and heat.
RUO – Research Use Only

General References:

  1. Hongguang Dai; Peter R.Chang; Fengying Geng; Jiugao Yu; Xiaofei Ma. Preparation and properties of thermoplastic starch/montmorillonite nanocomposite using N-(2-Hydroxyethyl)formamide as a new additive. Journal of Polymers and the Environment. 2009, 17, (4),225-232
  2. Hongguang Dai; Peter R.Chang; Fei Peng; Jiugao Yu; Xiaofei Ma. N-(2-Hydroxyethyl)formamide as a new plasticizer for thermoplastic starch. Journal of Polymer Research. 2009, 16, (5),529-535