2,4,5-Trimethyl-3-pyridinol, 97%, Thermo Scientific Chemicals
2,4,5-Trimethyl-3-pyridinol, 97%, Thermo Scientific Chemicals
2,4,5-Trimethyl-3-pyridinol, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,4,5-Trimethyl-3-pyridinol, 97%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
H52357.03
also known as H52357-03
1 g
Catalog number H52357.03
also known as H52357-03
Price (USD)
277.65
Online Exclusive
308.00
Save 30.35 (10%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
277.65
Online Exclusive
308.00
Save 30.35 (10%)
Each
Specifications
Beilstein115543
CAS5622-78-6
Chemical Name or Material2,4,5-Trimethyl-3-pyridinol
Formula Weight137.18
Health Hazard 1H315-H319-H335
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only