4-Methoxyindole-2-carboxylic acid, 97+%
4-Methoxyindole-2-carboxylic acid, 97+%
4-Methoxyindole-2-carboxylic acid, 97+%
4-Methoxyindole-2-carboxylic acid, 97+%
Thermo Scientific Chemicals

4-Methoxyindole-2-carboxylic acid, 97+%

CAS: 103260-65-7 | C10H9NO3 | 191.186 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
5 g
Catalog number H52392.03
also known as H52392-03
Price (USD)
130.65
Special offer
Online exclusive
Ends: 30-Jun-2026
153.00
Save 22.35 (15%)
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)
130.65
Special offer
Online exclusive
Ends: 30-Jun-2026
153.00
Save 22.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS103260-65-7
IUPAC Name4-methoxy-1H-indole-2-carboxylic acid
Molecular FormulaC10H9NO3
InChI KeyZZAVIQXQBBOHBB-UHFFFAOYSA-N
SMILESCOC1=C2C=C(NC2=CC=C1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormSolid
Assay from Supplier's CofA≥97.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only