Specifications
CAS332-48-9
Chemical Name or Material1-(2-Bromoethoxy)-4-fluorobenzene
Formula Weight219.05
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
View more