1-(2-Bromoethoxy)-4-fluorobenzene, 97+%
1-(2-Bromoethoxy)-4-fluorobenzene, 97+%
1-(2-Bromoethoxy)-4-fluorobenzene, 97+%
1-(2-Bromoethoxy)-4-fluorobenzene, 97+%
Thermo Scientific Chemicals

1-(2-Bromoethoxy)-4-fluorobenzene, 97+%

CAS: 332-48-9 | C8H8BrFO | 219.05 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H52403.06
also known as H52403-06
Price (USD)
428.65
Online Exclusive
476.00
Save 47.35 (10%)
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
428.65
Online Exclusive
476.00
Save 47.35 (10%)
Each
Add to cart
Specifications
CAS332-48-9
Chemical Name or Material1-(2-Bromoethoxy)-4-fluorobenzene
Formula Weight219.05
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only