Specifications
CAS674792-06-4
Chemical Name or Material(S)-1-Boc-2-isobutylpiperazine
Formula Weight242.36
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
View more